Drum faucets
Leverancier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
08540 08910
119-0236EA 68.3 EUR
119-0236 119-0240
Drum faucets
Kranen en gootstenen Kranen
These faucets can be simply placed on your drum for leakproof dispensing of flammable liquids. The brass construction with a polytetrafluoroethylene (PTFE) seal ensures excellent chemical resistance.
  • Safe and easy dispensing
  • FM approved
The faucets feature an internal flame arrester that prevents flashback of fire into the drum for added safety.