Safety containers for corrosives/acids, self-close
Leverancier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12161 12260 12762 12765
JUSU12161EA 76.3 EUR
JUSU12161 JUSU12260 JUSU12762 119-0273
Safety containers for corrosives/acids, self-close
Veiligheidscontainers
Self-close corrosive containers with stainless steel hardware are specifically designed to resist acids and caustics. Oval containers make the most of storage capacity and are marked in liters, quarts and gallons to permit accurate filling and dispensing.
HDPE, translucent.
  • Spring-close cap provides a tight seal
  • Flame arrester-free spout allows quick pouring of non-flammable corrosives