Base materials
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RC-109 RC-110 RC-117 RC-116 RC-111
RC-109EA 91.1 EUR
RC-109 RC-110 RC-117 RC-116 RC-111
Base materials
Media voor microbiologie Basismaterialen voor microbiologie
Tryptone is an assortment of peptides generated by the tryptic digestion of casein. It is commonly used in microbiology to produce Lysogeny broth for the growth of E. coli and other microorganisms, as it provides a source of amino acids.
  • Available in different pack sizes