Wipes for polishing applications, VeraClean Polish Plus
Leverancier: PGI NONWOVEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
74101 74104
115-2500EA 28.9 EUR
115-2500 115-2501
Wipes for polishing applications, VeraClean Polish Plus
Schoonmaakdoekjes Herbruikbare reinigingsdoeken Doeken voor algemeen gebruik
VeraClean Polish Plus is a bulky, velvet-soft polishing cloth that is designed for use in critical environments and on sensitive surfaces. It is suitable for use with almost all industrial polishing pastes. Due to its dense structure, VeraClean Polish Plus provides an alternative to high viscosity polish material.
  • High absorbency allows easy removal of excess polishing paste, water, oil and ink
  • Extremely soft, making it ideal for polishing
  • Resistant to most solvents and chemicals
  • Solvent-, silicone- and chemical-free
  • Low linting
Ideal for maintenance and repair of office equipment and for gold and silver processing. Also suitable for use in electronics and computer industries, laboratories, healthcare and in printing and associated industries.