Dishes, crystallising
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4210403 4210409 4210411 4210413 4210500 4210507 4210514
MARI4210403EA 47.7 EUR
MARI4210403 MARI4210409 MARI4210411 MARI4210413 MARI4210500 MARI4210507 MARI4210514
Dishes, crystallising
Reservoirs/Schalen/Potten Kristalliseerschalen
Crystallising dishes, made of borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance up to +500 °C
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Cylindrical shape with fused rim and flat bottom.