NMR tubes, PRECISION, with screw cap, SP Wilmad-LabGlass
Leverancier: SP Industries
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
535-TR-7 513-7TRA-7 335-TR-7 507-TR-7 507-TR-8 528-TR-7 528-TR-8 535-TR-8
WILM535-TR-7EA 43.8 EUR
WILM535-TR-7 WILM513-7TRA-7 WILM335-TR-7 WILM507-TR-7 WILM507-TR-8 WILM528-TR-7 WILM528-TR-8 WILM535-TR-8
NMR tubes, PRECISION, with screw cap, SP Wilmad-LabGlass
Buisjes NMR-buisjes
Borosilicate glass, Class A, ASTM Type 1.
These tubes are commonly used in sample degasification these tubes can maintain vacuum quality of >10⁻⁴
Leveringsinformatie: Tube comes complete with open top screw cap and septum.