Test sieves, 300×75 mm
Leverancier: ENDECOTTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
300SIW.050 300SIW1.00 300SIW.100 300SIW.800 300SIW.850 300SIW2.00 300SIW2.80 300SIW3.35 300SIW.280
ENDE300SIW.050EA 535 EUR
ENDE300SIW.050 ENDE300SIW1.00 ENDE300SIW.100 ENDE300SIW.800 ENDE300SIW.850 ENDE300SIW2.00 ENDE300SIW2.80 ENDE300SIW3.35 ENDE300SIW.280
Test sieves, 300×75 mm
Zeven Testzeven
Stainless steel woven wire mesh test sieves are ideal for all types of laboratory sampling and particle size analysis.
For more size options please contact your local Avantor office.
Certificaten: ISO3310-1:2000, BS410-1:2000; DIN-ISO3310-1; NF ISO3310-1:2000.
Leveringsinformatie: Each sieve is supplied together with a certificate of examination and carries the BS 410 kite mark.