Safety shoes, lace-up, U-Manager, London
Leverancier: U-Power
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MG20024/38 MG20024/39 MG20024/40 MG20024/41 MG20024/42 MG20024/43 MG20024/44 MG20024/45 MG20024/46 MG20024/47
UPOWMG20024/38EA 78.6 EUR
UPOWMG20024/38 UPOWMG20024/39 UPOWMG20024/40 UPOWMG20024/41 UPOWMG20024/42 UPOWMG20024/43 UPOWMG20024/44 UPOWMG20024/45 UPOWMG20024/46 UPOWMG20024/47
Safety shoes, lace-up, U-Manager, London
Schoenen en laarzen
Metal-free safety shoes made from soft full-grain leather and a slip-resistant, hard wearing PU sole.
  • Micro-Tech fibre
  • Executive plus anatomic and breathable insole with micro- cell for always cool and fresh feet
  • Natural fit width 11 for additional comfort
  • Airtoe protective Composite toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Certificaten: EN 20345:2011; S1, SRC