Magnetic screw caps, ND18 and septum
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
702827 702055 702072 702136
MANA702827EA 72.8 EUR
MANA702827 MANA702055 MANA702072 MANA702136
Magnetic screw caps, ND18 and septum
Sluitingen Sluitingen voor chromatografievials
The magnetic screw closures are ready assembled with septa suitable for headspace GC.
  • Magnetic srew caps with or without center hole
  • Flat surface of cap for a safe grip at the magnet
  • Thinner septum (1,5 instead of 3 mm) for a precise penetration of the needle (especially for SPME)
  • Septa made from high quality silicone/PTFE (also slitted), Butyl/PTFE or Red Rubber/TEF