Safety shoes, lace-up, SK Grip, Gessato Grip
Leverancier: U-Power
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RI20074/35 RI20074/36 RI20074/37 RI20074/38 RI20074/39 RI20074/40 RI20074/41 RI20074/42 RI20074/43 RI20074/44 RI20074/45 RI20074/46 RI20074/47
UPOWRI20074/35EA 97.8 EUR
UPOWRI20074/35 UPOWRI20074/36 UPOWRI20074/37 UPOWRI20074/38 UPOWRI20074/39 UPOWRI20074/40 UPOWRI20074/41 UPOWRI20074/42 UPOWRI20074/43 UPOWRI20074/44 UPOWRI20074/45 UPOWRI20074/46 UPOWRI20074/47
Safety shoes, lace-up, SK Grip, Gessato Grip
Schoenen en laarzen
Safety shoes made from water repellent smooth nubuck leather with and a lightweight, extremely flexible, slip resistant PU/PU U-Grip sole.
  • Anatomically shaped Polysoft insole made from soft PU, with pockets in the foot support area
  • PU/PU walking sole with 90° angle blocks and mulitlevel stabalising channels to for extremely high anti-slip features; TPU anti twist insert for additional support
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Airtoe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Certificaten: EN 20345:2011; S3, SRC