VWR®, Standards for water determination by the Karl Fischer method
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
85482.005EA 196 EUR
85482.005 85483.002 85483.005 85483.020 85484.002 85484.005 85484.020 85485.002 85485.005 85485.020 85487.004 85488.230 85489.260 85692.100 85449.100 85487.008 88427.004 88427.008 88428.001 88428.100 88429.008 88432.100 88433.005 88434.004 88435.004 85488.260 85488.290
VWR®, Standards for water determination by the Karl Fischer method
Standaarden Standaarden voor waterbepaling (door de Karl Fischer methode)
The tightening of regulatory requirements and the increasing demands of QA/QC methodology is becoming more important across all market sectors. The demand for standards to support testing methods continues to increase, and this growth applies to water determination according to Karl Fischer. Avantor now offers a complete range of Karl Fischer water standards as CRMs to calibrate and monitor equipment and check test results.
  • CRMs acc. to ISO 17025 / ISO 17034
  • CRM acc. to ASTM (in liquid petroleum)
  • Broad product range for both volumetric + KF coulometric titrations and indirect oven method
  • Liquid and solid water standards available
  • Supplied with a detailed Certificate of Analysis
Our range of Karl Fischer water standards are Certified Reference Materials acc to ISO 17025 and ISO 17034. The standards are available both in liquid and solid form.
We also offer a range of Karl Fischer water standards as CRM for determination in liquid petroleum according to ASTM. The water content in standards in liquid petroleum is determined using the ASTM D6304 method.
Certificaten: The water standards in liquid petroleum conform to ASTM D1744, E1064, D4377
Verpakking: When the Cat. No. (Pk) ends with a
0.005 then it is 10×5 ml
0.002 then it is 10×2 ml
0.020 then it is 5×20 ml