NMR tube washer, SP Wilmad-LabGlass
Leverancier: SP Industries
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WILMWG-1207-J10EA 223 EUR
WILMWG-1207-J10
NMR tube washer, SP Wilmad-LabGlass
Buisjes NMR-buisjes
NMR tube washer is fitted to a filter flask and a inverted sample tube is inserted into the washer, the solvent is introduced into the reservoir using a series of wash bottles. Can perform numerous wash steps and finish by pulling air through the tube.