Safety ankle boots, lace-up, Style & Job, Fuchs UK
Leverancier: U-Power
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
UA10614/35 UA10614/36 UA10614/37 UA10614/38 UA10614/39 UA10614/40 UA10614/41 UA10614/42 UA10614/43 UA10614/44 UA10614/45 UA10614/46 UA10614/47 UA10614/48
114-3161EA 95.3 EUR
114-3161 114-3162 114-3163 114-3164 114-3165 114-3166 114-3167 114-3168 114-3169 114-3170 114-3171 114-3172 114-3173 114-3174
Safety ankle boots, lace-up, Style & Job, Fuchs UK
Schoenen en laarzen
Anti-static safety ankle boots made from water resistant leather with a PU scuff cap and a slip-resistant and hard wearing PU sole.
  • Light non-slip action dry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Air toe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex'covers 100% of the sole of the foot
Colour: Black
Colour: Black
Certificaten: EN 20345:2011; S3, SRC