Tank liners, HyPerforma™
Leverancier: THERMO BIOPROCESS DIVISION
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SH30647.04 SH30647.01 SH30647.05 SH30647.02 SH30647.03 SH30646.01 SH30646.02 SH30646.03
HYCPSH30647.04EA 1920 EUR
HYCPSH30647.04 HYCPSH30647.01 HYCPSH30647.05 HYCPSH30647.02 HYCPSH30647.03 HYCPSH30646.01 HYCPSH30646.02 HYCPSH30646.03
Tank liners, HyPerforma™
Zakken Mediazakken
Tank liners are gamma irradiated, open single use bags designed to fit in cylindrical drums.
  • Gusset design allows for a close fit to the tank
  • Constructed of CX3-9 film
  • For HyPerforma™ Single-Use Mixer DS 300