Safety ankle boots, lace-up, Rock & Roll, Florida
Leverancier: U-Power
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RR10356/38 RR10356/39 RR10356/40 RR10356/41 RR10356/42 RR10356/43 RR10356/44 RR10356/45 RR10356/46 RR10356/47
UPOWRR10356/38EA 94.3 EUR
UPOWRR10356/38 UPOWRR10356/39 UPOWRR10356/40 UPOWRR10356/41 UPOWRR10356/42 UPOWRR10356/43 UPOWRR10356/44 UPOWRR10356/45 UPOWRR10356/46 UPOWRR10356/47
Safety ankle boots, lace-up, Rock & Roll, Florida
Schoenen en laarzen
100% metal-free safety ankle boots made from soft nubuck-tech with highly breathable textile mesh inserts and a slip-resistant dual-density PU sole.
  • Comfortable, anatomically formed insole ergo dry features a dual PE/EVA support system with textile surface; anti-static and anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Airtoe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Grey/black
Colour: Grey/black
Certificaten: EN 20345:2011; S1P, SRC