Safety boots, lace-up, Rock & Roll, Ghepard UK
Leverancier: U-Power
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RR70324/44 RR70324/43
UPOWRR70324/44EA 111 EUR
UPOWRR70324/44 UPOWRR70324/43
Safety boots, lace-up, Rock & Roll, Ghepard UK
Schoenen en laarzen
100% metal-free safety boots made from smooth pull-up water repellent leather with PU scuff and a slip-resistant dual-density PU sole.
  • Comfortable, anatomically formed insole ergo dry features a dual PE/EVA support system with textile surface; anti-static and anti-fungal treated
  • Warm fur lining
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Air toe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Colour: Black
Certificaten: EN 20345:2011; S3, CI, SRC