Stalagmometers, SP Wilmad-LabGlass
Leverancier: SP Industries
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LG-5050-100 LG-5050-102 LG-5050-104 LG-5050-106
WILMLG-5050-100EA 1870 EUR
WILMLG-5050-100 WILMLG-5050-102 WILMLG-5050-104 WILMLG-5050-106
Stalagmometers, SP Wilmad-LabGlass
stalagmometers
Stalagmometers designed for checking the surface tension of liquids.
  • Accurate calibration
  • Drop count method
  • Graduation above and below the bulb
Designed for checking the surface tension of liquids.
Leveringsinformatie: The set is supplied with 2,5, 3,5 and 5 ml tubes in a protective case.