Single channel pipettes, fixed volume
Leverancier: Vitrex Medical A/S
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
560013 561013 562013 564013
MODU560013EA 34.4 EUR
MODU560013 MODU561013 MODU562013 MODU564013
Single channel pipettes, fixed volume
Pipetten Micropipetten
This miniature fixed volume pipette is specially designed for use with Diagnostic Kits. At a length of 130 mm it is the ideal size for blood collecting.
  • Two step plunger operation
  • Tip cone designed to accept most VITREX® capillary tubes
  • Fully autoclavable at 121 °C and 15 psi
  • Colour-coded for easy identification
  • Each pipette is individually calibrated as per the ISO 8655 standard