Safety ankle boots, lace-up, U-Spider, Venus, Shuttle
Leverancier: U-Power
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SP10014/39 SP10014/40 SP10014/41 SP10014/42 SP10014/43 SP10014/44 SP10014/45 SP10014/46 SP10014/47
UPOWSP10014/39EA 105.2 EUR
UPOWSP10014/39 UPOWSP10014/40 UPOWSP10014/41 UPOWSP10014/42 UPOWSP10014/43 UPOWSP10014/44 UPOWSP10014/45 UPOWSP10014/46 UPOWSP10014/47
Safety ankle boots, lace-up, U-Spider, Venus, Shuttle
Schoenen en laarzen
100% metal-free safety ankle boots made from extremely soft, abrasion resistant, water repellent leather and a heat resistant to 300 °C, antislip nitrile rubber sole.
  • Light non-slip action dry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Innovative polyester collar
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Air toe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Colour: Black
Certificaten: EN 20345:2011; S3, SRC, HRO