Chlorine reagent set
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2440400 2459500
HACH2440400EA 73.4 EUR
HACH2440400 HACH2459500
Chlorine reagent set
Milieutestkits
This chlorine reagent set has 100 tests each to test chlorine (free and total) by DPD colorimetic method.