Glass tubing
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
246180180 246300200 246400320 246420160 246440160 246650220 246090150
KAVA246180180EA 376 EUR
KAVA246180180 KAVA246300200 KAVA246400320 KAVA246420160 KAVA246440160 KAVA246650220 KAVA246090150
Glass tubing
Slangen Transfer Tubing
Borosilicate glass, SIMAX.
  • Very good chemical resistance
  • Very good temperature resistance