Test tubes
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A620025001221 A61001200101 A61601600121 A61801800121 A62003000121 A61001200102 A61601600122 A610016001211
SCERA620025001221EA 2880 EUR
SCERA620025001221 SCERA61001200101 SCERA61601600121 SCERA61801800121 SCERA62003000121 SCERA61001200102 SCERA61601600122 SCERA610016001211
Test tubes
Buisjes Reageerbuisjes
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For a wide variety of applications.