Erlenmeyer flasks, narrow neck
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
94206001 94206005
SCER94206001EA 38.9 EUR
SCER94206001 SCER94206005
Erlenmeyer flasks, narrow neck
Kolven Erlenmeyers
Borosilicate glass 3,3, KIMAX®.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With graduation.
Certificaten: DIN ISO 1773