Tubing kits
Leverancier: GRANT INSTRUMENTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HOSE100 HOSE200
GRANHOSE100EA 311 EUR
GRANHOSE100 GRANHOSE200
Tubing kits
Slangen Transfer Tubing
Tubing kit 2×2 m, assembled with OptimaTM pump outlet plate and simple hose clips, no tools required (can be cut to length as required).