Sterilin™ Samples Containers with Screw Caps, PS, 60 to 250 ml
Leverancier: STERLIN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
125AM 125CM 165A 165B 190A 125AP 185AM 125BM 125CP 125BP 165C 185AP 190C 185BM 185CM 185BP 185CP 190A/1 190B 190B/1 190A/IRR 190AIW 190C/IRR 125AMWC
215-0315EA 199 EUR
215-0315 215-0319 215-0356 215-0357 215-1772 215-0316 215-1765 215-0317 215-0320 215-0318 215-0358 215-1764 215-1792 215-1757 215-1755 215-1756 215-1729 SLIN190A/1 215-1779 SLIN190B/1 SLIN190A/IRR SLIN190AIW SLIN190C/IRR SLIN125AMWC
Sterilin™ Samples Containers with Screw Caps, PS, 60 to 250 ml
Monsternamepotjes Containers voor algemeen gebruik
Contain samples in Thermo Scientific™ Sterilin™ Polystyrene containers with 60 to 250 ml capacities.
  • Aseptically manufactured
  • For in vitro use only
  • 215-0315, 215-0317 and 215-0319 product certified 95kPa-compliant ensuring suitability for sample transport
Available with either plastic or metal flow seal caps.
Certificaten: Leak-tested in accordance with BS EN 14254 Annexe D. CE marked in accordance with the European Directive 98/79/EC. 125AM, 125BM and 125CM product certified 95kPa-compliant ensuring suitability for sample transport.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904