Sterilin™ Containers, Bijou, with Screw Cap
Leverancier: STERLIN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
129B 129A 129BBAC M00525
215-0329EA 285 EUR
215-0329 215-0328 216-2250 SLINM00525
Sterilin™ Containers, Bijou, with Screw Cap
Monsternamepotjes Containers voor algemeen gebruik
Polystyrene, clear, with polyethylene screw cap. Store small-volume samples in Thermo Scientific™ Sterilin™ 7 ml Polystyrene Bijou Containers. Ideal for pediatric samples and other small-volume clinical applications. Aseptically manufactured. Not available in America.
  • Available pre-filled with boric acid (0,09 g) for the preservation of urine samples
  • Suitable for centrifugation at 7200×g
  • Aseptically manufactured
  • Available with glass coverslip, ideal for work on the culture of intracellular organisms (eg. Chlamydia trachomatis, Rickettsia etc)
  • In vitro use only
Disposable plastic bijou container available with or without label. Containers are ideal for small volume samples.
Certificaten: Leak-tested in accordance with BS EN 14254 Annexe D. CE marked in accordance with the European Directive 98/79/EC.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904