Moisturising hand and body lotion, Stokolan® Light PURE
Leverancier: Deb-STOKO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RES100ML RES1L
115-2331EA 46.68 EUR
115-2331 115-2139
Moisturising hand and body lotion, Stokolan® Light PURE
Huidverzorging Huidcrèmes Hand- en bodylotions
Softcream (O/W emulsion) with moisturising effect for the effective care of normal skin. Suitable for all over body use and for use under latex and nitrile gloves. Suitable for people prone to allergies and eczema. Ideal for use in industrial, food handling and healthcare work areas, office and public washrooms.
  • Contains glycerine, allantoin and shea butter to moisturise and condition the skin
  • Non-tainting to food
  • Colourant-free, silicone-free, perfume-free
Certificaten: ECARF Quality Seal for allergy friendly products, HACCP
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904