Lactodensimeters according to Quevenne
Leverancier: Ludwig Schneider GmbH & CO. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3302331 3302341 3302341-20
SCNL3302331EA 19.5 EUR
SCNL3302331 SCNL3302341 SCNL3302341-20
Lactodensimeters according to Quevenne
Hydrometers
Lactodensimeters calibrated for special applications.
Error limit: ±0,001 sp/gr
Bestelinformatie: A works or DAkkS calibration certificate is available on request.
Verpakking: In square plastic sleeve.