Chlorophosphonazo indicator solution for hardness determination
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2589599 2589549
HACH2589599EA 413 EUR
HACH2589599 HACH2589549
Chlorophosphonazo indicator solution for hardness determination
Milieutestkits
For the determination of ultra low range hardness by chlorophosphonazo method. Ideal for use with Pour-Thru cell. Requires CDTA reagent.