Immersion oil
Leverancier: ZEISS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4449600000000
4449640000000
630-0340EA
52
EUR
630-0340
ZEIS4449640000000
Immersion oil
Kleurstoffen
Microscopy and staining reagents
Immersion oil 518 F according ISO 8036-1/2.
- Halogen-free
- Fluorescence free
Certificaten: ISO 8036-1/2