Tris(ethylenediamine)rhodium(III) chloride trihydrate (23,2% Rh)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
10553.02 10553.03
10553.02EA 344 EUR
10553.02 10553.03
Tris(ethylenediamine)rhodium(III) chloride trihydrate (23,2% Rh)
Tris(ethylenediamine)rhodium(III) chloride trihydrate
CAS No.: 15004-86-1 Molecular Formula: C6H24Cl3N6Rh.3H2O Formula Weight: 443.61 (389.56anhy) Physical Form: Crystalline/Fused lump, crystals fuse over time Appearance: White Melting Point: 280° dec. MDL No.: MFCD00149661
Formule: C₆H₃₀Cl₃N₆O₃Rh
MW: 443.601 g/mol
Smeltpunt: 280 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149661
CAS nummer: 15004-86-1
EINECS: 237-846-7