Ammonium hexachloroiridate(III) hydrate ≥99.99% (metals basis), Premion®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
10713.03
10713.06
10713.03EA
343
EUR
10713.03
10713.06
Ammonium hexachloroiridate(III) hydrate ≥99.99% (metals basis), Premion®
Ammonium hexachloroiridate(III) hydrate
Purity: 99.99%
CAS No.: 15752-05-3
Molecular Formula: (NH4)3IrCl6.xH2O
Formula Weight: 459.06(anhy)
Physical Form: Crystalline powder
Sensitivity: Hygroscopic
Appearance: Green
Odour: Odourless
MDL No.: MFCD00003393
Formule:
(NH₄)₃IrCl₆·xH₂O Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003393 CAS nummer: 15752-05-3 EINECS: 239-842-0 |