Iron(II) perchlorate hydrate, Reagent Grade
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11646.22
11646.36
11646.22EA
44.6
EUR
11646.22
11646.36
Iron(II) perchlorate hydrate, Reagent Grade
Iron(II) perchlorate hydrate
CAS No.: 335159-18-7
Molecular Formula: Fe(ClO4)2.xH2O
Formula Weight: 254.75(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Melting Point:
Formule:
Fe(ClO₄)₂·xH₂O |
MDL Nummer:
MFCD00150234 CAS nummer: 335159-18-7 |