Dihydrogen dinitrosulphatoplatinate(II) solution (4 - 6% Pt)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

12627.NA 12627.NC 12627.NJ
12627.NAEA 308 EUR
12627.NA 12627.NC 12627.NJ
Dihydrogen dinitrosulphatoplatinate(II) solution (4 - 6% Pt)
Dihydrogen dinitrosulphatoplatinate(II)

CAS No.: 12033-81-7 Molecular Formula: H2Pt(NO2)2SO4 Formula Weight: 384.17(anhy) Physical Form: Liquid UN No.: UN3264 MDL No.: MFCD00064668

Formule: H₂Pt(NO₂)₂SO₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064668
CAS nummer: 12033-81-7
UN: 3264
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR