Silver two part conductive adhesive, EXP. XX/XX resin and hardener
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12942.03
12942.09
12942.03EA
53.6
EUR
12942.03
12942.09
Silver two part conductive adhesive, EXP. XX/XX resin and hardener
Silver two part conductive adhesive, EXP. XX/XX resin and hardener
A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite
Expiration date provided
Limited shelf life. 60 days after shipment.
MDL No.: MFCD00003397
Applications: A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite