Stainless steel (Fe:Cr:Ni; 73:18:9 wt%), rods, Ø 3.175 mm (0.125 in) Type 303
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13475.BV
13475.RM
13475.BVEA
31.1
EUR
13475.BV
13475.RM
Stainless steel (Fe:Cr:Ni; 73:18:9 wt%), rods, Ø 3.175 mm (0.125 in) Type 303
Stainless steel
Molecular Formula: Fe:Cr:Ni; 73:18:9 wt%
MDL No.: MFCD00148499