Neodymium boron iron, magnet alloy additive Nd:B:Fe (15:8:77) atomic%, REacton®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13642.18
13642.30
13642.95
13642.18EA
96.3
EUR
13642.18
13642.30
13642.95
Neodymium boron iron, magnet alloy additive Nd:B:Fe (15:8:77) atomic%, REacton®
Neodymium boron iron, magnet alloy additive
Molecular Formula: Nd:B:Fe; 15:8:77 atomic%; Magnet Alloy Additive
Physical Form: Broken Lump
Odour: Odourless
MDL No.: MFCD02091729