Scandium(III) carbonate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13646.03 13646.06
13646.03EA 112 EUR
13646.03 13646.06
Scandium(III) carbonate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Scandium(III) carbonate hydrate
Purity: 99.99% Molecular Formula: Sc2(CO3)3.xH2O Formula Weight: 269.94(anhy) Physical Form: Crystalline Melting Point: 500° dec. MDL No.: MFCD00011501
Formule: Sc₂(CO₃)₃·xH₂O
MW: 269.94 g/mol (anhy)
Smeltpunt: >500 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011501
CAS nummer: 5809-49-4