Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), soluble -325 mesh, Thickness: 0.4-1.2 µm Type 316-L
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13787.30
13787.A1
13787.30EA
49.2
EUR
13787.30
13787.A1
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), soluble -325 mesh, Thickness: 0.4-1.2 µm Type 316-L
Stainless steel
Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
MDL No.: MFCD00148499