Telluric acid dihydrate ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

14197.09 14197.18
14197.09EA 96.7 EUR
14197.09 14197.18
Telluric acid dihydrate ≥99%
Telluric acid dihydrate

Purity: 99+% CAS No.: 13451-14-4 Molecular Formula: H2O4Te.2H2O Formula Weight: 229.64 (193.61anhy) Physical Form: Crystalline Density: 3.158 Sensitivity: Hygroscopic Appearance: White Melting Point: 136° Boiling Point: 160° dec. UN No.: UN3284 MDL No.: MFCD00150387

Formule: H₂O₄Te.2H₂0
Kookpunt: 160 °C (ontbinding)
Smeltpunt: 136 °C
Dichtheid: 3,158
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150387
CAS nummer: 13451-14-4
EINECS: 232-267-6
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR