Samarium(III) acetate hydrate ≥99.9%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14604.14
14604.22
14604.14EA
105
EUR
14604.14
14604.22
Samarium(III) acetate hydrate ≥99.9%
Samarium(III) acetate hydrate
Purity: 99.9%
CAS No.: 17829-86-6
Molecular Formula: Sm(OOCCH3)3.xH2O
Formula Weight: 327.48(anhy)
Physical Form: Crystalline Aggregates
Density: 1.94
Sensitivity: Hygroscopic
Odour: Odourless
Boiling Point: 300°
Solubility: Soluble in water
MDL No.: MFCD00150124
Formule:
Sm(OOCCH₃)₃·xH₂O Kookpunt: 300 °C Dichtheid: 1,94 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00150124 CAS nummer: 17829-86-6 EINECS: 233-950-1 |