Heavy Metals, plasma standard solution, Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100µg/ml, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14657.AE
14657.AP
14657.AEEA
189
EUR
14657.AE
14657.AP
Heavy Metals, plasma standard solution, Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100µg/ml, Specpure®
Standaarden
Kationische standaarden
Standaarden met meerdere elementen
UN No.: UN3264
MDL No.: MFCD02091713