Copper(II) gluconate
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Copper(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic Acid Copper(II) Salt, Copper(II) D-gluconate, Copper Gluconate
15325.18 15325.30 15325.A1
15325.18EA 38 EUR
15325.18 15325.30 15325.A1
Copper(II) gluconate
Copper(II) gluconate
CAS No.: 527-09-3 Molecular Formula: [HOCH2[CH(OH)]4CO2]2Cu Formula Weight: 453.85 Physical Form: Powder Melting Point: 155-157° MDL No.: MFCD00075297
Formule: C₁₂H₂₂CuO₁₄
MW: 453.84 g/mol
Smeltpunt: 155…157 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00075297
CAS nummer: 527-09-3
EINECS: 208-408-2