Cobalt(II) tetrafluoroborate hydrate ≥96%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
23142.14
23142.22
23142.14EA
30
EUR
23142.14
23142.22
Cobalt(II) tetrafluoroborate hydrate ≥96%
Cobalt(II) tetrafluoroborate hydrate
Purity: 96%
CAS No.: 26490-63-1
Molecular Formula: Co(BF4)2.xH2O
Formula Weight: 232.54(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
UN No.: UN3260
MDL No.: MFCD00016012
Formule:
Co(BF₄)₂·xH₂O Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00016012 CAS nummer: 26490-63-1 ADR: 8,III |