Tricarbonylnitrosylcobalt
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
23151.03
23151.06
23151.03EA
69.7
EUR
23151.03
23151.06
Tricarbonylnitrosylcobalt
Tricarbonylnitrosylcobalt
CAS No.: 14096-82-3
Molecular Formula: C3CoNO4
Formula Weight: 172.97
Physical Form: Liquid
Density: 1.47
Sensitivity: Air Sensitive
Appearance: Dark red
Odour: Unpleasant
Melting Point: -1°
Boiling Point: 48.6°(55° dec.)
UN No.: UN1992
MDL No.: MFCD00016014
Formule:
C₃CoNO₄ Kookpunt: 48,6 °C/ 55 °C (kook./ dec.) Smeltpunt: –1 °C Dichtheid: 1,47 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00016014 CAS nummer: 14096-82-3 EINECS: 237-945-5 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...