Ceramic tubes, one end closed
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30093.DR 30093.DW
30093.DREA 246 EUR
30093.DR 30093.DW
Ceramic tubes, one end closed
Slangen Transfer Tubing
Pure Al₂O₃ Aluminium oxide AL24.
Slightly porous, good resistance to thermal shock, extremely good creep strength.