Ceramic tubes, both ends open
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31606.EH 31606.EO 31609.EH
31606.EHEA 277 EUR
31606.EH 31606.EO 31609.EH
Ceramic tubes, both ends open
Slangen Transfer Tubing
Pure Al₂O₃ Aluminium oxide AL24.
Slightly porous, good resistance to thermal shock, extremely good creep strength.