Ceramic discs, round, AL23, with hole
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
33140.KF 33147.KF
33140.KFEA 57.2 EUR
33140.KF 33147.KF
Ceramic discs, round, AL23, with hole
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.