Combustion trays, ceramic, AL23
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
33183.KF 33185.KF 33186.KF
33183.KFEA 80.9 EUR
33183.KF 33185.KF 33186.KF
Combustion trays, ceramic, AL23
Schuitjes Verbrandingsschuitjes
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.