Gold tin eutectic ≥99.99% (metals basis), foil, Thickness: 0.1 mm (0.004 in) x 43.8 mm (1.72 in) wide
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
36310.BS
36310.BW
36310.BSEA
1388
EUR
36310.BS
36310.BW
Gold tin eutectic ≥99.99% (metals basis), foil, Thickness: 0.1 mm (0.004 in) x 43.8 mm (1.72 in) wide
Gold tin eutectic
Purity: 99.99%
Molecular Formula: Au:Sn; 80:20 wt%
Physical Form: ≈6.37g/10cm
MDL No.: MFCD02091709